Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SSB Antibody (OTI2C8), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274356H
Description
SSB Monoclonal specifically detects SSB in Human, Canine, Primate, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
SSB | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
SSB | |
Full-length protein expressed in 293T cell transfected with human SSB expression vector. | |
0.1 mL | |
Primary | |
Human, Canine, Primate, Monkey | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI2C8 | |
Western Blot, Immunohistochemistry | |
La autoantigen, La ribonucleoprotein, lupus La protein, member 3, Sjoegren syndrome type B antigen, Sjogren syndrome antigen B (autoantigen La), SS-B, SS-B/La protein | |
Mouse | |
Affinity Purified | |
Core ESC Like Genes, Stem Cell Markers | |
6741.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction