Learn More
Invitrogen™ SSB Polyclonal Antibody

Description
Positive test controls include: Jurkat, LO2, HepG2, 293T. The target is usually found in the following locations: Nucleus. Immunogen sequence: KFVETPGQKY KETDLLILFK DDYFAKKNEE RKQNKVEAKL RAKQEQEAKQ KLEEDAEMKS LEEKIGCLLK FSGDLDDQTC REDLHILFSN HGEIKWIDFV RGAKEGIILF KEKAKEALGK AKDANNGNLQ LRNKEVTWEV LEGEVEKEAL KKIIEDQQES LNKWKSKGRR FKGKGKGNKA AQPGSGKGKV QFQGKKTKFA SDDEHDEHDE NGATGPVKRA REETDKEEPA SKQQKTENGA GDQ.
Specifications
Specifications
| Antigen | SSB |
| Applications | ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 4.89 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | SSB |
| Gene Accession No. | P05455 |
| Gene Alias | autoantigen La; La; la autoantigen; La autoantigen homolog; La protein; la ribonucleoprotein; La ribonucleoprotein domain family, member 3; La/SSB; LARP3; lupus La antigen; lupus La protein; Lupus La protein homolog; Lupus LA protein homolog (LA ribonucleoprotein) (LA autoantigen homolog); lupus La protein-like; mRNA for autoantigen; OTTHUMP00000204777; sjoegren syndrome type B antigen; Sjogren syndrome antigen B; Sjogren syndrome antigen B (autoantigen La); small RNA binding exonuclease protection factor La; Ssb; SS-B; SS-B/La protein |
| Gene Symbols | SSB |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.