Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ SSB Polyclonal Antibody
Supplier: Thermo Scientific™ PA5109337
Description

Specifications
SSB | |
Polyclonal | |
Unconjugated | |
SSB | |
autoantigen La; La; la autoantigen; La autoantigen homolog; La protein; la ribonucleoprotein; La ribonucleoprotein domain family, member 3; La/SSB; LARP3; lupus La antigen; lupus La protein; Lupus La protein homolog; Lupus LA protein homolog (LA ribonucleoprotein) (LA autoantigen homolog); lupus La protein-like; mRNA for autoantigen; OTTHUMP00000204777; sjoegren syndrome type B antigen; Sjogren syndrome antigen B; Sjogren syndrome antigen B (autoantigen La); small RNA binding exonuclease protection factor La; Ssb; SS-B; SS-B/La protein | |
Rabbit | |
Affinity Chromatography | |
RUO | |
6741 | |
-20°C, Avoid Freeze/Thaw Cycles, store in dark | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
P05455 | |
SSB | |
Recombinant protein within Human SSB aa 165-346. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction