Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SSBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117079
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117079 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117079 Supplier Invitrogen™ Supplier No. PA5117079
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HL-60, MCF7, SW620 Immunogen sequence: MFRRPVLQVL RQFVRHESET TTSLVLERSL NRVHLLGRVG QDPVLRQVEG KNPVTIFSLA TNEMWRSGDS EVYQLGDVSQ KTTWHRISVF RPGLRDVAYQ YVKKGSRIYL EGKIDYGEYM DKNNVRRQAT TIIADNIIFL SDQTKEKE.

This protein binds preferentially and cooperatively to ss-DNA. Probably involved in mitochondrial DNA replication. SSBP is a housekeeping gene involved in mitochondrial biogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SSBP1
Applications ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.52 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Ssbp1
Gene Accession No. Q04837, Q9CYR0
Gene Alias 2810480P10Rik; G630031O20Rik; mtDBP; mtSSB; Mt-SSB; PWP1-interacting protein 17; single strand DNA-binding protein P16; single stranded DNA binding protein 1; single-stranded DNA binding protein 1; single-stranded DNA binding protein 1, mitochondrial; single-stranded DNA-binding protein, mitochondrial; SOSS-B1; Ssbp; Ssbp1
Gene Symbols Ssbp1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-148 of human SSBP1 (NP_0012434421).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 381760, 6742
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.