Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SSBP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537065
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537065 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537065 Supplier Invitrogen™ Supplier No. PA537065
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 38 kDa. Purity is >95% by SDS-PAGE.

SSBP2 is a member of a closely related, evolutionarily conserved, and ubiquitously expressed gene family, potential tumor suppressor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SSBP2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene SSBP2
Gene Accession No. P81877, Q9CYZ8
Gene Alias 1500004K09Rik; 2310079I02Rik; 9330163K02Rik; A830008M03Rik; AU067692; AW558684; DKFZp686F03273; HSPC116; RGD1562968; Sequence-specific single-stranded-DNA-binding protein 2; si:ch211-219m24.4; single stranded DNA binding protein 2; single-stranded DNA binding protein 2; single-stranded DNA-binding protein 2; SOSS-B2; Ssbp2; Ssdp2
Gene Symbols SSBP2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 2-47 of human SSBP2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23635, 361877, 66970
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.