Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SSBP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540431
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540431 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540431 Supplier Invitrogen™ Supplier No. PA540431
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: YPGGPRPPLR IPNQALGGVP GSQPLLPSGM DPTRQQGHPN MGGPMQRMTP Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 92%.

SSBP2 is a member of a closely related, evolutionarily conserved, and ubiquitously expressed gene family, potential tumor suppressor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SSBP2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SSBP2
Gene Accession No. P81877
Gene Alias 1500004K09Rik; 2310079I02Rik; 9330163K02Rik; A830008M03Rik; AU067692; AW558684; DKFZp686F03273; HSPC116; RGD1562968; Sequence-specific single-stranded-DNA-binding protein 2; si:ch211-219m24.4; single stranded DNA binding protein 2; single-stranded DNA binding protein 2; single-stranded DNA-binding protein 2; SOSS-B2; Ssbp2; Ssdp2
Gene Symbols SSBP2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human SSBP2 (aa 131-180).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23635
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.