Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SSBP3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA530487
Description
Recommended positive controls: Jurkat, Raji. Predicted reactivity: Mouse (88%), Rat (86%), Xenopus laevis (84%), Chicken (98%), Rhesus Monkey (88%), Bovine (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
May be involved in transcription regulation of the alpha 2(I) collagen gene where it binds to the single-stranded polypyrimidine sequences in the promoter region.
Specifications
| SSBP3 | |
| Polyclonal | |
| Unconjugated | |
| SSBP3 | |
| 2610021L12Rik; 2610200M23Rik; 5730488C10Rik; AI854733; AW551939; CSDP; LAST; Lck-associated signal transducer; sequence-specific single-stranded-DNA-binding protein; single stranded DNA binding protein 3; single-stranded DNA binding protein 3; single-stranded DNA-binding protein 3; Ssbp3; SSDP; SSDP1; Ssdp3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23648 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.66 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9BWW4 | |
| SSBP3 | |
| Recombinant protein encompassing a sequence within the center region of human SSBP3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction