Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SSBP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544323
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544323 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544323 Supplier Invitrogen™ Supplier No. PA544323
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SNFPMGPGSD GPMGGMGGME PHHMNGSLGS GDIDGLPKNS PNNISGISNP Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.

May be involved in transcription regulation of the alpha 2(I) collagen gene where it binds to the single-stranded polypyrimidine sequences in the promoter region.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SSBP3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SSBP3
Gene Accession No. Q9BWW4
Gene Alias 2610021L12Rik; 2610200M23Rik; 5730488C10Rik; AI854733; AW551939; CSDP; LAST; Lck-associated signal transducer; sequence-specific single-stranded-DNA-binding protein; single stranded DNA binding protein 3; single-stranded DNA binding protein 3; single-stranded DNA-binding protein 3; Ssbp3; SSDP; SSDP1; Ssdp3
Gene Symbols SSBP3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human SSBP3.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23648
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.