Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SSBP3 Polyclonal Antibody

SSBP3 Polyclonal Antibody
Supplier: Thermo Scientific PA5107280
Description
SSBP3 Polyclonal Antibody for Western Blot
Specifications
| SSBP3 | |
| Polyclonal | |
| Unconjugated | |
| SSBP3 | |
| 2610021L12Rik; 2610200M23Rik; 5730488C10Rik; AI854733; AW551939; CSDP; LAST; Lck-associated signal transducer; sequence-specific single-stranded-DNA-binding protein; single stranded DNA binding protein 3; single-stranded DNA binding protein 3; single-stranded DNA-binding protein 3; Ssbp3; SSDP; SSDP1; Ssdp3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 23648 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| Q9BWW4 | |
| SSBP3 | |
| Synthetic peptide directed towards the middle region of human SSBP3 (aa 339-388). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction