Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals SSC (20X), RNAse free

$127.20 - $228.00

Specifications

Chemical Name or Material SSC
Concentration 20X
Physical Form Liquid
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ60561AP
View Documents
Thermo Scientific Chemicals
J60561AP
500 mL
Each for $228.00
Order in Bulk

Request bulk or custom formats

Only null left
Add to Cart
 
AAJ60561AK
View Documents
Thermo Scientific Chemicals
J60561AK
250 mL
Each for $127.20
Order in Bulk

Request bulk or custom formats

Only null left
Add to Cart
 
Description

Description

SSC solution is commonly used in nucleic acid hybridization techniques at concentrations ranging from 0.1X to 20X depending on the application.  It is used to facilitate transfer of nucleic acids to membranes in Southern and Northern blotting, dot blotting, and colony and plaque lifts. 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
SSC solution is commonly used in nucleic acid hybridization techniques at concentrations ranging from 0.1X to 20X depending on the application.  It is used to facilitate transfer of nucleic acids to membranes in Southern and Northern blotting, dot blotting, and colony and plaque lifts. 

Solubility
Soluble in water.

Notes
0.3M sodium citrate, 3.0M sodium chloride, pH 7.0.
Specifications

Specifications

SSC
20X
Liquid
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.