Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Molecular Grade 20X (pH 7.0) SSC Buffer

Catalog No. PRV4261 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1000mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PR-V4261 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PR-V4261 Supplier Promega Supplier No. V4261
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Used in nucleic acid hybridization techniques at concentrations from 0.1X to 20X.

  • Quality tested and certified free of DNase, RNase and protease activity

Specifications

Chemical Name or Material SSC Buffer 20X
Physical Form Liquid
Quantity 1000mL
Grade Molecular Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.