Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SSC5D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559819
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559819 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559819 Supplier Invitrogen™ Supplier No. PA559819
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TCTGYTDYDD YPPWTWDPTS REDLAKGTTT AGVPGHTLPW RTTRRPGSSS PAIRRLPDTG SKDGYKLPWT WDTPSGRGLA EGTPTAGKLG PTL Highest antigen sequence identity to the following orthologs: Mouse - 65%, Rat - 61%.

Binds to extracellular matrix proteins. Binds to pathogen-associated molecular patterns (PAMPs) present on the cell walls of Gram-positive and Gram-negative bacteria and fungi, behaving as a pattern recognition receptor (PRR). Induces bacterial and fungal aggregation and subsequent inhibition of PAMP-induced cytokine release. Does not possess intrinsic bactericidal activity. May play a role in the innate defense and homeostasis of certain epithelial surfaces.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SSC5D
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SSC5D
Gene Accession No. A1L4H1
Gene Alias S5D-SRCRB; scavenger receptor cysteine rich domain containing (5 domains); scavenger receptor cysteine rich family member with 5 domains; scavenger receptor cysteine rich family, 5 domains; scavenger receptor cysteine-rich glycoprotein; soluble scavenger protein with 5 SRCR domains; Soluble scavenger receptor cysteine-rich domain-containing protein SSC5D; soluble scavenger with 5 domains; SSC5D
Gene Symbols SSC5D
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SSC5D. Recombinant protein control fragment (Product #RP-99201).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284297
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.