Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SSDD/Steroid sulfatase Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985706
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5706 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5706 Supplier Bioss Supplier No. BS3857R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene catalyzes the conversion of sulfated steroid precursors to estrogens during pregnancy. The encoded protein is found in the endoplasmic reticulum, where it acts as a homodimer. Mutations in this gene are known to cause X-linked ichthyosis (XLI).

Specifications

Antigen SSDD/Steroid sulfatase
Applications ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Sts
Gene Accession No. P08842
Gene Alias ArsC; ARSC1; Arylsulfatase C; ASC; ES; Estrone sulfatase; LOC402795 protein; si:ch211-271l9.1; SSDD; Steroid sulfatase; steroid sulfatase (microsomal), arylsulfatase C, isozyme S; steroid sulfatase (microsomal), isozyme S; steroid sulphatase; Steryl-sulfatase; Steryl-sulfate sulfohydrolase; STS; XLI
Gene Symbols Sts
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Steroid sulfatase.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 412
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.