Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SSEA-1 Antibody (6B11B4) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | CD15/Lewis X |
---|---|
Clone | 6B11B4 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000 |
Applications | Western Blot, ELISA |
Description
SSEA-1 Monoclonal specifically detects SSEA-1 in Human samples. It is validated for Western Blot, ELISA.Specifications
CD15/Lewis X | |
1 mg/mL | |
Western Blot, ELISA | |
Unconjugated | |
RUO | |
Human | |
CD 15, CD-15, Lewis X, LeX, SSEA-1, stage-specific embryonic antigen 1 | |
FUT4 | |
IgG2b | |
Affinity Purified |
6B11B4 | |
Western Blot 1:500 - 1:2000, ELISA 1:10000 | |
Monoclonal | |
Mouse | |
Cellular Markers, Embryonic Stem Cell Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Immunology, Pancreatic Stem Cell Markers, Stem Cell Markers | |
PBS with 0.05% Sodium Azide | |
2526 | |
Purified recombinant fragment of human FUT4 (AA: 199-302) expressed in E. Coli. | |
Primary | |
59 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title