Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SSEA-1 Antibody (CL6334), Novus Biologicals™

Mouse Monoclonal Antibody
$382.00 - $610.00
Specifications
Antigen | SSEA-1 |
---|---|
Clone | CL6334 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
SSEA-1 Monoclonal specifically detects SSEA-1 in Human samples. It is validated for Western Blot.Specifications
SSEA-1 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
alpha-(1,3)-fucosyltransferase, CD15, EC 2.4.1, EC 2.4.1.-, EC 2.4.1.65, ELAM-1 ligand fucosyltransferase, ELFT, FCT3AELAM ligand fucosyltransferase, Fucosyltransferase 4, fucosyltransferase 4 (alpha (1,3) fucosyltransferase, myeloid-specific), Fucosyltransferase IV, fucT-IV, FUC-TIV, FUTIV, Galactoside 3-L-fucosyltransferase, Lewis X, LeX, SSEA-1, stage-specific embryonic antigen 1 | |
FUT4 | |
IgG2a | |
Protein A purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
CL6334 | |
Monoclonal | |
Purified | |
Cellular Markers, Embryonic Stem Cell Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Immunology, Pancreatic Stem Cell Markers, Stem Cell Markers, Stem Cells | |
PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide | |
2526.0 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: RKPSGAGWEKEWAEAPQEA | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title