Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SSEA-3 Antibody (MC-631) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB014035 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB014035 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB014035 Supplier Novus Biologicals Supplier No. NBP280962
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

SSEA-3 Monoclonal specifically detects SSEA-3 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, CyTOF-ready.

Specifications

Antigen SSEA-3
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MC-631
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500, Flow Cytometry : 10-20 μg/mL, Immunohistochemistry : 1:10-1:500, Immunocytochemistry/Immunofluorescence : 1:50-1:100, Immunoprecipitation : 1:10-1:500, Immunohistochemistry-Paraffin : 1:10-1:500, Immunohistochemistry-Frozen : 1:10-1:500, CyTOF-ready
Formulation PBS
Gene Alias Stage specific embryonic antigen 3, Stage-specific embryonic antigen 3
Host Species Rat
Immunogen 4-8 cell stage mouse embryos.
Purification Method IgM purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody recognizes the SSEA3 that is expressed upon the surface of human teratocarcinoma stem cells (EC), human embryonic germ cells (EG) and human embryonic stem cells (ES). No immunoreactivity is evident with undifferentiated murine EC, ES and EG cells. Expression of SSEA3 is down regulated following differentiation of human EC cells. In contrast, the differentiation of murine EC and ES cells may be accompanied by an increase in SSEA-3 expression.
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.