Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SSEA-4 Antibody (MC-813-70), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB1435Z
Description
SSEA-4 Monoclonal antibody specifically detects SSEA-4 in Human, Mouse samples. It is validated for Flow Cytometry, ImmunocytochemistrySpecifications
SSEA-4 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Human, Mouse | |
Purified |
Flow Cytometry, Immunocytochemistry | |
MC-813-70 | |
Flow Cytometry, Immunocytochemistry | |
SSEA-4,stage-specific embryonic antigen 4 | |
2120Ep human embryonal carcinoma cell line | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction