Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SSEA-4 Mouse anti-Canine, Chicken, Human, Mouse, Primate, Rabbit, DyLight 350, Clone: MC-813-70, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP226644UV0.1ML
Description
SSEA-4 Monoclonal specifically detects SSEA-4 in Human, Mouse, Canine, Chicken, Primate, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Flow (Cell Surface).Specifications
| SSEA-4 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Human, Mouse, Canine, Chicken, Primate, Rabbit | |
| Purified |
| Flow Cytometry | |
| MC-813-70 | |
| Flow Cytometry, Flow (Cell Surface) | |
| NP_803131 | |
| Human embryonal carcinoma cell line 2102Ep | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG3 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction