Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SSEA-4 Antibody (MC-813-70), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP226644AF594
Description
SSEA-4 Monoclonal specifically detects SSEA-4 in Human, Mouse, Canine, Chicken, Primate, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Flow (Cell Surface).Specifications
SSEA-4 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
Human embryonal carcinoma cell line 2102Ep | |
0.1 mL | |
Human, Mouse, Canine, Chicken, Primate, Rabbit | |
IgG3 κ |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
MC-813-70 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Flow (Cell Surface) | |
Mouse | |
Protein G purified | |
Primary | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction