Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SSEA-5 Mouse anti-Human, BV510, Clone: 8e11/SSEA-5 (also known as 8e11), BD Biosciences™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 750808
Description
SSEA-5 Monoclonal antibody specifically detects SSEA-5 in Human samples. It is validated for Flow Cytometry
Specifications
SSEA-5 | |
Monoclonal | |
Brilliant Violet 510 | |
Stage-Specific Embryonic Antigen-5 | |
Human Undifferentiated Embryonic Stem Cells | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
8e11/SSEA-5 (also known as 8e11) | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BV510 under optimal conditions that minimize unconjugated dye and antibody. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction