Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SSEA1 (CD15) Polyclonal Antibody, Abbomax

Catalog No. 502204494
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4494 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4494 Supplier Abbomax Inc Supplier No. 620470
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: HL-60; K560 Cellular location: Membrane Cytoplasm.

SSEA1 (CD15) Polyclonal antibody specifically detects SSEA1 (CD15) in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen SSEA1 (CD15)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Host Species Rabbit
Immunogen The Synthetic peptide corresponding to the C-term of human SSEA1 protein
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.