Learn More
Invitrogen™ SSEA1 Monoclonal Antibody (MEM-158), PE
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA119743
Description
This antibody reacts with CD15, a cell membrane molecule 3-fucosyl-N-acetyllactosamine (3-FAL) strongly expressed on the surface of granulocytes, monocytes, macrophages, mast cells; it is also present on Langerhans cells and some myeloid precursors cells. This antibody will not cross-react with porcine, bovine or sheep.
CD15 (Lewis X, Le(x); stage specific embryonic antigen-1, SSEA-1) is a trisacharide determinant (3-fucosyl-N-acetyllactosamine) expressed on several glycolipids, glycoproteins and proteoglycans of various cell types, e.g. granulocytes, mast cells, monocytes, macrophages, cells of gastric mucosa, nervous system or various tumour cells. There are several variants of Lewis x, such as sialyl-Lewis x or sulphated Lewis x. Cells with high surface expression of Le(x) antigen exhibit strong self-aggregation, based on calcium-dependent Le(x)-Le(x) interaction. This process is involved for example in embryo compaction or in autoaggregation of teratocarcinoma cells. Sialyl-Le(x) and its isomer sialyl-Le(a) are ligands of selectins. CD15 expression has been extensively used to confirm diagnosis of Hodgkin's disease.
Specifications
SSEA1 | |
Monoclonal | |
PE | |
FUT4 | |
FUTIV; Lewis X; SSEA-1 | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
2526 | |
4°C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
MEM-158 | |
TBS with 0.2% BSA and 15mM sodium azide; pH 8 | |
P22083 | |
FUT4 | |
Human granulocytes. | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgM |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.