Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SSEA3 Monoclonal Antibody (MC-631)

Rat Monoclonal Antibody

Supplier:  Invitrogen™ MA1020X

 View more versions of this product

Catalog No. PIMA1020X


Only null left
Edge
Add to Cart

Description

Description

SSEA3 Monoclonal Antibody for ICC/IF, Flow

SSEA-3 (stage-specific embryonic antigen 3) is a cell surface antigen, expressed along with SSEA-4, TRA-1-60 and TRA-1-81 in embryonic stem cells, embryonal carcinoma cells, and induced pluripotent stem cells (iPS). In human cells, these surface markers are down-regulated during the differentiation process, while SSEA1 is absent in undifferentiated human stem cells and upregulated on the cell surface after retinoic acid mediated differentiation. Conversely, in mouse stem cells, SSEA-1 expression decreases with differentiation while SSEA-3 and SSEA-4 expression increase.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SSEA3
Monoclonal
1 mg/mL
Potassium Phosphate with 1 mg/mL BSA, 0.15M NaCl and 0.05% sodium azide
Rat
Affinity chromatography - MBP
RUO
Human
Antibody
IgM
Flow Cytometry, Immunocytochemistry
MC-631
Unconjugated
SSEA-3; stage-specific embryonic antigen 3
Fischer rat immunized with 4-8 cell stage mouse embryos.
20 μL
Primary
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.