Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ B3GALT5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103651
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103651 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103651 Supplier Invitrogen™ Supplier No. PA5103651
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total B3GALT5.

B3GALT5 catalyzes the transfer of Gal to GlcNAc-based acceptors with a preference for the core3 O-linked glycan GlcNAc(beta1,3)GalNAc structure. Can use glycolipid LC3Cer as an efficient acceptor. (Uniprot).
TRUSTED_SUSTAINABILITY

Specifications

Antigen B3GALT5
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene B3GALT5
Gene Accession No. Q9Y2C3
Gene Alias 1190002B21Rik; AU045265; B3GALT5; b3Gal-T5; b3Galt-V; B3GalTx; B3gt5; B3T5; Beta-1,3-galactosyltransferase 5; beta-1,3-GalTase 5; beta-3-galactosyltransferase 5; beta3GalT5; beta3Gal-T5; beta-3-Gx-T5; GLCT5; homolog of C. elegans Bt toxin resistance gene bre-5; SSEA-3 synthase; Stage-specific embryonic antigen 3 synthase; UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase 5; UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase, polypeptide 5; UDP-Gal:beta-GlcNAc beta-1,3-galactosyltransferase 5; UDP-galactose:beta-N-acetylglucosamine beta-1,3-galactosyltransferase 5
Gene Symbols B3GALT5
Host Species Rabbit
Immunogen A synthesized peptide derived from human B3GALT5(Accession Q9Y2C3), corresponding to amino acid residues M85-T135.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10317
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.