Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SSEA4 Monoclonal Antibody (MC-813-70)
GREENER_CHOICE

Catalog No. PIMA1021
Change view
Click to view available options
Quantity:
100 μg
20 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA1021 100 μg
PIMA1021X 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA1021 Supplier Invitrogen™ Supplier No. MA1021
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

SSEA4 Monoclonal Antibody for Flow, ICC/IF

SSEA-4 (Stage-specific embryonic antigen 4) is a cell surface antigen, expressed along with SSEA-3, TRA-1-60 and TRA-1-81 in embryonic stem cells, embryonal carcinoma cells and induced pluripotent stem cells (iPS). In human cells, these surface markers are down-regulated during the differentiation process, while SSEA-1 is absent in undifferentiated human stem cells and upregulated on the cell surface after retinoic acid mediated differentiation. Conversely, in mouse stem cells, SSEA-1 expression decreases with differentiation while SSEA-3 and SSEA-4 expression increase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SSEA4
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone MC-813-70
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Potassium phosphate with 0.15M NaCl, 1mg/mL BSA and 0.05% sodium azide
Gene Alias SSEA-4; stage-specific embryonic antigen 4
Host Species Mouse
Immunogen human embryonal carcinoma cell line 2102Ep.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.