Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SSEA5 Monoclonal Antibody (8E11), PE
GREENER_CHOICE

Catalog No. PIMA1144PE
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA1144PE 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA1144PE Supplier Invitrogen™ Supplier No. MA1144PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunofluorescence and flow cytometric analysis of MA1-144-PE shows positive staining of SSEA-5 in human embryonal carcinoma NCCIT cells.

Stage-Specific Embryonic Antigen-5 (SSEA-5) is an oligosaccharide that can be part of glycosphingolipids and glycoproteins. It contains the motif Fuc1-2Gal-beta1-3GlcNAc-beta, which is characteristic of H-1 antigens. SSEA-5 is found in the inner cell mass of the human blastocyst during embryogenesis, on undifferentiated human embryonic stem cells and on the surface of induced pluripotent cells. SSEA-5 was found to proceed the loss of other known stem cell markers and the combination of SSEA-5 with either CD9 and CD90 or CD50 and CD200 was superior in identifying and eliminating rare undifferentiated human ES cells from differentiated cell populations than the use of other established stem cell markers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SSEA5
Applications Flow Cytometry
Classification Monoclonal
Clone 8E11
Concentration 1 mg/mL
Conjugate PE
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene Alias SSEA-5
Host Species Mouse
Immunogen Undifferentiated H9 human embryonic stem cells.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.