Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SSEA5 Monoclonal Antibody (8E11-SSEA5), eBioscience™
GREENER_CHOICE

Catalog No. 14885782
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-885-782 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-885-782 Supplier Invitrogen™ Supplier No. 14885782
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Description: The monoclonal antibody 8E11-SSEA5 recognizes human stage-specific embryonic antigen-5 (SSEA-5), an H type-1 glycan. SSEA-5 belongs to the SSEA family of glycoproteins and is highly expressed on pluripotent human stem cells including embryonic stem (ES) cells and induced pluripotent stem (iPS) cells. During embryonic development, SSEA-5 is expressed within the blastocyst on inner cell mass (ICM) cells that later give rise to ES cells. As ES and iPS cells differentiate, SSEA-5 expression is downregulated. The tumorigenic potential of contaminating stem cells in cell populations for transplantation is a hurdle to using this approach for disease therapy. The 8E11 antibody in combination with either CD9 and CD90 or CD50 and CD200 to sort populations has been shown to effectively separate teratoma-forming cells from stem cell-derived populations of differentiated cells. Applications Reported: This 8E11-SSEA5 antibody has been reported for use in flow cytometric analysis, western blotting, and immunocytochemistry. Applications Tested: This 8E11-SSEA5 antibody has been tested immunocytochemistry on unfixed human Ntera2 cells at less than or equal to 10 μg/mL. The 8E11-SSEA5 antibody has also been tested by flow cytometric analysis on NTERA-2 cells at less than or equal to 0.25 μg/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less...

Stage-Specific Embryonic Antigen-5 (SSEA-5) is an oligosaccharide that can be part of glycosphingolipids and glycoproteins. It contains the motif Fuc1-2Gal-beta1-3GlcNAc-beta, which is characteristic of H-1 antigens. SSEA-5 is found in the inner cell mass of the human blastocyst during embryogenesis, on undifferentiated human embryonic stem cells and on the surface of induced pluripotent cells. SSEA-5 was found to proceed the loss of other known stem cell markers and the combination of SSEA-5 with either CD9 and CD90 or CD50 and CD200 was superior in identifying and eliminating rare undifferentiated human ES cells from differentiated cell populations than the use of other established stem cell markers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SSEA5
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 8E11-SSEA5
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Alias SSEA-5
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.