Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SSFA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565127
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565127 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565127 Supplier Invitrogen™ Supplier No. PA565127
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HIIEITEVEED LFPAETVELL REASAESDVG KSSESEFTQY TTHHILKSLA SIEAKCSDMS SENTTGPPSS MDRVNTALQR AQMKVCSLSN QRMG Highest antigen sequence identity to the following orthologs - mouse 67%, rat 64%.

SSFA2 gene ontology annotations related to this gene include actin binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SSFA2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ITPRID2
Gene Accession No. P28290
Gene Alias 5730488C15Rik; cleavage signal-1 protein; CS1; CS-1; ITPRID2; ITPR-interacting domain-containing protein 2; KIAA1927; Ki-ras-induced actin-interacting protein; Krap; KRAS-induced actin-interacting protein; mKIAA1927; Protein ITPRID2; SPAG13; sperm associated antigen 13; sperm specific antigen 2; sperm-specific antigen 2; Sperm-specific antigen 2 homolog; SSFA2
Gene Symbols ITPRID2
Host Species Rabbit
Immunogen Recombinant Human SSFA2. Recombinant protein control fragment (Product #RP-104648).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6744
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.