Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SSFA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100275
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100275 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100275 Supplier Invitrogen™ Supplier No. PA5100275
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SSFA2.

SSFA2 gene ontology annotations related to this gene include actin binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SSFA2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ITPRID2
Gene Accession No. P28290, Q922B9
Gene Alias 5730488C15Rik; cleavage signal-1 protein; CS1; CS-1; ITPRID2; ITPR-interacting domain-containing protein 2; KIAA1927; Ki-ras-induced actin-interacting protein; Krap; KRAS-induced actin-interacting protein; mKIAA1927; Protein ITPRID2; SPAG13; sperm associated antigen 13; sperm specific antigen 2; sperm-specific antigen 2; Sperm-specific antigen 2 homolog; SSFA2
Gene Symbols ITPRID2
Host Species Rabbit
Immunogen A synthesized peptide derived from human SSFA2(Accession P28290), corresponding to amino acid residues L1033-S1083.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 311146, 6744, 70599
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.