Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SSH1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP323410
Description
SSH1 Polyclonal antibody specifically detects SSH1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
SSH1 | |
Polyclonal | |
Western Blot 1:1000, Immunohistochemistry 1:100, Immunocytochemistry/ Immunofluorescence 1:250 | |
EC 3.1.3.16, EC 3.1.3.48, FLJ21928, hSSH-1L, KIAA1298FLJ38102, protein phosphatase Slingshot homolog 1, slingshot 1, slingshot homolog 1 (Drosophila), SSH1L, SSH-1L, SSH-like protein 1 | |
A synthetic peptide (coupled to KLH) corresponding to amino acid residues in the C-terminal region of human SSH1. This sequence has 100% homology with similar regions of mouse and bovine SSH1, and has low homology to SSH2L and SSH3L. [UniProt# Q8WYL5] | |
0.1 mL | |
Phospho Specific | |
54434 | |
Store at -20°C. | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
PBS, 1 mg/mL BSA and 50% Glycerol | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction