Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SSH1 Mouse anti-Human, Clone:1A5C8, Abnova™

Catalog No. 89333110 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-110 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-110 Supplier Abnova Corporation Supplier No. MAB16660
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of human SSH1.

The ADF (actin-depolymerizing factor)/cofilin family (see MIM 601442) is composed of stimulus-responsive mediators of actin dynamics. ADF/cofilin proteins are inactivated by kinases such as LIM domain kinase-1 (LIMK1; MIM 601329). The SSH family appears to play a role in actin dynamics by reactivating ADF/cofilin proteins in vivo (Niwa et al., 2002 [PubMed 11832213]).[supplied by OMIM]

Specifications

Antigen SSH1
Applications ELISA, Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 1A5C8
Conjugate Unconjugated
Description slingshot homolog 1 (Drosophila)
Dilution ELISA (1:10000)
Western Blot
Immunohistochemistry
Immunocytochemistry (1:200-1:1000)
Flow Cytometry (1:200-1:400)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene SSH1
Gene Alias FLJ38102, KIAA1298, SSH-1
Host Species Mouse
Immunogen A synthetic peptide corresponding to amino acid 1032-1044 of human SSH1 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 54434
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.