Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SSH1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5143765
Description
The antibody detects a 150 kDa* protein corresponding to the molecular mass of SSH1L on SDS-PAGE immunoblots of PC12 cells.
This gene encodes a member of the SWI/SNF family, whose members have helicase and ATPase activities and are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein is part of the large ATP-dependent chromatin remodeling complex SNF/SWI, which is required for transcriptional activation of genes normally repressed by chromatin. It possesses at least two conserved domains that could be important for its function. First, it has a DNA-binding domain that can specifically bind an AT-rich DNA sequence known to be recognized by a SNF/SWI complex at the beta-globin locus. Second, the C-terminus of the protein can stimulate glucocorticoid receptor-dependent transcriptional activation. It is thought that the protein encoded by this gene confers specificity to the SNF/SWI complex and may recruit the complex to its targets through either protein-DNA or protein-protein interactions. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
SSH1 | |
Polyclonal | |
Unconjugated | |
SSH1 | |
hSSH-1L; KIAA1298; Protein phosphatase Slingshot homolog 1; slingshot homolog 1; slingshot protein phosphatase 1; SSH1; SSH1L; SSH-1L; SSH-like protein 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
231637, 304580, 54434 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide | |
Q76I79, Q8WYL5 | |
SSH1 | |
A synthetic peptide (coupled to KLH) corresponding to amino acid residues in the C-terminal region of human SSH1L. This sequence has 100% homology with similar regions of mouse and bovine SSH1L, and has low homology to SSH2L and SSH3L. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction