Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SSH3 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10060674V
Description
SSH3 Polyclonal antibody specifically detects SSH3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
SSH3 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
EC 3.1.3.16, EC 3.1.3.48, FLJ10928, FLJ20515, FLJ42240, hSSH-3L, protein phosphatase Slingshot homolog 3, slingshot 3, slingshot homolog 3 (Drosophila), SSH3, SSH3L, SSH-3L, SSH-like protein 3 | |
The immunogen recognized by this antibody maps to a region between residue 550 and 600 of human slingshot homolog 3 using the numbering given in entry NP_060327.2 (GeneID 54961). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
54961 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction