Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SSH3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101788
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101788 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101788 Supplier Invitrogen™ Supplier No. PA5101788
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SSH3.

The ADF (actin-depolymerizing factor)/cofilin family is composed of stimulus-responsive mediators of actin dynamics. ADF/cofilin proteins are inactivated by kinases such as LIM domain kinase-1 (LIMK1). The SSH family appears to play a role in actin dynamics by reactivating ADF/cofilin proteins in vivo.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SSH3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Ssh3
Gene Accession No. Q5XIS1, Q8K330, Q8TE77
Gene Alias BC028922; hSSH-3L; mSSH-3L; Protein phosphatase Slingshot homolog 3; slingshot 3; slingshot homolog 3; slingshot homolog 3 (Drosophila); slingshot protein phosphatase 3; slingshot-3L; slingshot-like 3; Ssh3; SSH-3; Ssh3l; SSH-3L; SSH-like protein 3
Gene Symbols Ssh3
Host Species Rabbit
Immunogen A synthesized peptide derived from human SSH3(Accession Q8TE77), corresponding to amino acid residues T356-G406.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 245857, 365396, 54961
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.