Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SSM 3 trifluoroacetate
GREENER_CHOICESDP

Catalog No. 525310 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
525310 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 525310 Supplier Tocris Bioscience™ Supplier No. 5253/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent furin inhibitor

SSM 3 trifluoroacetate is a potent furin inhibitor (EC50 = 54 nM). Blocks furin-dependent cell surface processing of anthrax protective antigen-83 in vitro.

Specifications

Chemical Name or Material rel-N,N′′-[[(1R,3S,4S,6R)-4,6-Bis[(aminoiminomethyl)amino]-1,3-cyclohexanedyil]bis(oxy-4,1-phenylene)]bisguanidine tetratrifluoroacetate salt
CAS 922732-52-3
Quantity 10 mg
Target Additional Synthetic Protease Inhibitors
Molecular Formula C22H32N12O2.4CF3CO2H
Formula Weight 952.66
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.