Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SSMEM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144166
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144166 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144166 Supplier Invitrogen™ Supplier No. PA5144166
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SSMEM1 Polyclonal Antibody for ICC/IF, IHC (P), ELISA

Chromosome 7 has been linked to Osteogenesis imperfecta, Pendred syndrome, Lissencephaly, Citrullinemia and Shwachman-Diamond syndrome. The deletion of a portion of the q arm of chromosome 7 is associated with Williams-Beuren syndrome, a condition characterized by mild mental retardation, an unusual comfort and friendliness with strangers and an elfin appearance. Deletions of portions of the q arm of chromosome 7 are also seen in a number of myeloid disorders including cases of acute myelogenous leukemia and myelodysplasia. The C7orf45 gene product has been provisionally designated C7orf45 pending further characterization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SSMEM1
Applications ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene SSMEM1
Gene Accession No. Q8WWF3
Gene Alias 1700016K02Rik; 1700025E21Rik; C7orf45; serine rich single-pass membrane protein 1; serine-rich single-pass membrane protein 1; SSMEM1
Gene Symbols SSMEM1
Host Species Rabbit
Immunogen Recombinant Human Serine-rich single-pass membrane protein 1 protein (56-244AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 136263
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.