Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ sspA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA580253
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA580253 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA580253 Supplier Invitrogen™ Supplier No. PA580253
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Escherichia coli ssPA/Stringent starvation protein A.

sspA preferentially cleaves peptide bonds on the carboxyl-terminal side of aspartate and glutamate. Along with other extracellular proteases it is involved in colonization and infection of human tissues. Required for proteolytic maturation of thiol protease SspB and inactivation of SspC, an inhibitor of SspB. It is the most important protease for degradation of fibronectin-binding protein (FnBP) and surface protein A, which are involved in adherence to host cells. May also protect bacteria against host defense mechanism by cleaving the immunoglobulin classes IgG, IgA and IgM. May be involved in the stability of secreted lipases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen sspA
Applications ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias Glutamyl endopeptidase; V8 protease; V8 proteinase
Host Species Rabbit
Immunogen Recombinant Escherichia coli ssPA/Stringent starvation protein A protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.