Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ssPA Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA580252
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for E. coli ssPA/Stringent starvation protein A.
sspA preferentially cleaves peptide bonds on the carboxyl-terminal side of aspartate and glutamate. Along with other extracellular proteases it is involved in colonization and infection of human tissues. Required for proteolytic maturation of thiol protease SspB and inactivation of SspC, an inhibitor of SspB. It is the most important protease for degradation of fibronectin-binding protein (FnBP) and surface protein A, which are involved in adherence to host cells. May also protect bacteria against host defense mechanism by cleaving the immunoglobulin classes IgG, IgA and IgM. May be involved in the stability of secreted lipases.
Specifications
ssPA | |
Polyclonal | |
Unconjugated | |
Glutamyl endopeptidase; V8 protease; V8 proteinase | |
Recombinant E. coli ssPA/Stringent starvation protein A protein. | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA | |
1 mg/mL | |
PBS with no preservative | |
Rabbit | |
Protein A | |
RUO | |
Bacteria | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction