Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ssPA/Stringent Starvation Protein A Antibody (113), DyLight 680, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290173FR
Description
ssPA/Stringent Starvation Protein A Monoclonal specifically detects ssPA/Stringent Starvation Protein A in E. coli samples. It is validated for ELISA.Specifications
ssPA/Stringent Starvation Protein A | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
944744 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
113 | |
ELISA | |
ECK3218, Pog, ssp, stringent starvation protein A | |
This antibody was obtained from a rabbit immunized with purified, recombinant E. coli ssPA/Stringent Starvation Protein A (P0ACA3; Met1-Ser212). | |
0.1 mL | |
Primary | |
E. coli | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction