Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ SSR 146977 hydrochloride
GREENER_CHOICESDP

Catalog No. 329750 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-975-0 50 mg
32-971-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 32-975-0 Supplier R&D Systems™ Supplier No. 3297/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective NK3 antagonist

Potent and selective NK3 receptor antagonist (Ki values are 0.26 and 19.3 nM in CHO cells expressing the human NK3 and NK2 receptor respectively). Also inhibits senktide -induced inositol monophosphate formation and intracellular calcium mobilization (IC50 = 10 nM).

Chemical Identifiers

CAS 264618-38-4
Molecular Formula C35H43Cl3N4O2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 658.11
Purity >98%
Molecular Weight (g/mol) 658.105
Chemical Name or Material SSR 146977 hydrochloride
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.