Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SSR 149415
GREENER_CHOICE

Catalog No. 619510
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-951-0 10 mg
61-955-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 61-951-0 Supplier Tocris Bioscience™ Supplier No. 6195/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective vasopressin V1B antagonist

SSR 149415 is a potent and selective vasopressin V1B antagonist (Ki values are 3.7 and 1.3 nM for native and recombinant rat V1b receptors, respectively). Displays >70-fold selectively for rat V1B over V1A, V2 oxytoxin and a range of other receptors and ion channels. Inhibits AVP-induced glucagon release in In-R1-G9 α-pancreatic cells in vitro. Inhibits stress-induced ACTH release and exhibits anxiolytic-like effects in anxiety models. Orally bioavailable.

Specifications

Chemical Name or Material (2S,4R)-1-[(3R)-5-Chloro-1-[(2,4-dimethoxyphenyl)sulfonyl]-2,3-dihydro-3-(2-methoxyphenyl)-2-oxo-1H-indol-3-yl]-4-hydroxy-N,N-dimethyl-2-pyrrolidinecarboxamide
CAS 439687-69-1
Assay Percent Range >98%
Quantity 10 mg
Molecular Formula C30H32ClN3O8S
Formula Weight Observed MW: 630.11
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.