Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SSR 180711 hydrochloride
GREENER_CHOICESDP

Catalog No. 555950 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-595-0 50 mg
55-591-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 55-595-0 Supplier Tocris Bioscience™ Supplier No. 5559/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Partial agonist at α7 nAChR

SSR 180711 hydrochloride is a partial agonist at α7 nAChR. Enhances episodic memory and reverses MK-801 -induced deficits in retention of episodic memory. Increases extracellular dopamine levels in rat prefrontal cortex. Also exhibits antidepressant-like properties.

Specifications

Chemical Name or Material 4-Bromophenyl 1,4-diazabicyclo[3.2.2]nonane-4-carboxylate hydrochloride
CAS 446031-79-4
Quantity 50 mg
Target Nicotinic (a7) Receptor Agonists
Molecular Formula C14H17BrN2O2.HCl
Formula Weight 361.66
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.