Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SSR1 Monoclonal Antibody (OTI 5D5), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA503265

Description
The signal sequence receptor (SSR) is a glycosylated endoplasmic reticulum (ER) membrane receptor associated with protein translocation across the ER membrane. The SSR consists of 2 subunits, a 34-kD glycoprotein encoded by this gene and a 22-kD glycoprotein. This gene generates several mRNA species as a result of complex alternative polyadenylation. This gene is unusual in that it utilizes arrays of polyA signal sequences that are exclusively non-canonical.Specifications
SSR1 | |
Monoclonal | |
0.29 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P43307 | |
SSR1 | |
Full length human recombit protein of human SSR1 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Western Blot | |
OTI 5D5 | |
Unconjugated | |
SSR1 | |
TRAPA | |
Mouse | |
Affinity Chromatography | |
RUO | |
6745 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction