Learn More
SSR1 Mouse anti-Human, Clone: OTI4H6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF503144

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The signal sequence receptor (SSR) is a glycosylated endoplasmic reticulum (ER) membrane receptor associated with protein translocation across the ER membrane. The SSR consists of 2 subunits, a 34-kD glycoprotein encoded by this gene and a 22-kD glycoprotein. This gene generates several mRNA species as a result of complex alternative polyadenylation. This gene is unusual in that it utilizes arrays of polyA signal sequences that are exclusively non-canonical.Specifications
SSR1 | |
Monoclonal | |
Unconjugated | |
SSR1 | |
TRAPA | |
Mouse | |
Affinity Chromatography | |
RUO | |
6745 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI4H6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P43307 | |
SSR1 | |
Full length human recombit protein of human SSR1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.