Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SSR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120727
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120727 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120727 Supplier Invitrogen™ Supplier No. PA5120727
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HepG2, A-549, SKOV3, Mouse liver, Mouse pancreas, Rat liver, Rat pancreas. Immunogen sequence: EEGARLLASK SLLNRYAVEG RDLTLQYNIY NVGSSAALDV ELSDDSFPPE DFGIVSGMLN VKWDRIAPAS NVSHTVVLRP LKAGYFNFTS ATITYLAQED GPVVIGSTSA PGQGGILAQR EFDRRFSP.

The signal sequence receptor (SSR) is a glycosylated endoplasmic reticulum (ER) membrane receptor associated with protein translocation across the ER membrane. The SSR consists of 2 subunits, a 34-kD glycoprotein (alpha-SSR or SSR1) and a 22-kD glycoprotein (beta-SSR or SSR2).The signal sequence receptor (SSR) is a glycosylated endoplasmic reticulum (ER) membrane receptor associated with protein translocation across the ER membrane. The SSR consists of 2 subunits, a 34-kD glycoprotein (alpha-SSR or SSR1) and a 22-kD glycoprotein (beta-SSR or SSR2). The human beta-signal sequence receptor gene (SSR2) maps to chromosome bands 1q21-q23.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SSR2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.60 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Ssr2
Gene Accession No. P43308, Q9CPW5
Gene Alias 1500032E05Rik; AI315033; AU020133; DKFZp686F19123; glycoprotein 25H; gp25H; HSD25; signal sequence receptor beta subunit; signal sequence receptor subunit 2; signal sequence receptor subunit beta; signal sequence receptor, beta; signal sequence receptor, beta (translocon-associated protein beta); signal sequence receptor, beta precursor; Ssr2; SSR2 protein; SSR-beta; TLAP; translocon-associated protein beta; translocon-associated protein subunit beta; translocon-associated protein subunit bet-like protein; TRAP[b]; TRAPb; TRAPbeta; TRAP-beta; wu:fb75b04; zgc:92180
Gene Symbols Ssr2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 18-145 of human SSR2 (NP_003136.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 295235, 66256, 6746
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.