Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SSR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542730
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542730 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542730 Supplier Invitrogen™ Supplier No. PA542730
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SFVVLALFAV TQAEEGARLL ASKSLLNRYA VEGRDLTLQY NIYNVGSSAA Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 100%.

The signal sequence receptor (SSR) is a glycosylated endoplasmic reticulum (ER) membrane receptor associated with protein translocation across the ER membrane. The SSR consists of 2 subunits, a 34-kD glycoprotein (alpha-SSR or SSR1) and a 22-kD glycoprotein (beta-SSR or SSR2).The signal sequence receptor (SSR) is a glycosylated endoplasmic reticulum (ER) membrane receptor associated with protein translocation across the ER membrane. The SSR consists of 2 subunits, a 34-kD glycoprotein (alpha-SSR or SSR1) and a 22-kD glycoprotein (beta-SSR or SSR2). The human beta-signal sequence receptor gene (SSR2) maps to chromosome bands 1q21-q23.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SSR2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Ssr2
Gene Accession No. P43308
Gene Alias 1500032E05Rik; AI315033; AU020133; DKFZp686F19123; glycoprotein 25H; gp25H; HSD25; signal sequence receptor beta subunit; signal sequence receptor subunit 2; signal sequence receptor subunit beta; signal sequence receptor, beta; signal sequence receptor, beta (translocon-associated protein beta); signal sequence receptor, beta precursor; Ssr2; SSR2 protein; SSR-beta; TLAP; translocon-associated protein beta; translocon-associated protein subunit beta; translocon-associated protein subunit bet-like protein; TRAP[b]; TRAPb; TRAPbeta; TRAP-beta; wu:fb75b04; zgc:92180
Gene Symbols Ssr2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human SSR2 (aa 5-54).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6746
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.