Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SSR3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599059
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599059 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599059 Supplier Invitrogen™ Supplier No. PA599059
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SSR3 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

SSR3 (signal sequence receptor, gamma) is also known as TRAPG. By immunoaffinity chromatography, biochemical purification, peptide sequence analysis, and screening of a rat liver cDNA library, Hartmann et al. (1993) obtained a cDNA encoding Ssr3, which they termed Trap-gamma. The deduced 185-amino acid protein spans the membrane 4 times and is expressed as a 20-kD protein. Gross (2012)mapped the SSR3 gene to chromosome 3q25.31 based on an alignment of the SSR3 sequence with the genomic sequence (GRCh37). Hartmann et al. (1993)found that Ssr3 was associated in equal amounts with Ssr1, Ssr2, and Ssr4 in canine pancreatic microsomes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SSR3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Ssr3
Gene Accession No. Q9UNL2
Gene Alias 0610038P07Rik; AL022999; AU022074; AW553833; signal sequence receptor subunit 3; signal sequence receptor subunit gamma; signal sequence receptor, gamma; signal sequence receptor, gamma (translocon-associated protein gamma); SSR gamma; Ssr3; SSR-gamma; translocon-associated protein gamma; translocon-associated protein gamma subunit; translocon-associated protein subunit gamma; TRAP-complex gamma subunit; TRAPG; TRAP-gamma
Gene Symbols Ssr3
Host Species Rabbit
Immunogen Recombinant Human Translocon-associated protein subunit gamma protein (77-135AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6747
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.