Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SSR4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560948
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560948 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560948 Supplier Invitrogen™ Supplier No. PA560948
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TPSYYTTSDA VISTETVFIV EISLTCKNRV QNMALYADVG GKQFPVTRGQ DVGRYQVSWS LDHKSAHAGT YEVRFFDEES YSLLRKAQRN NEDISIIPPL FTVSVDHRGT WNGPWVSTEV LAA Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

TRAP proteins are part of a complex whose function is to bind calcium to the ER membrane and thereby regulate the retention of ER resident proteins. SSR4. also called TRAPD. is assumed to be involved in protein secretion. It is located in the Xq28 region.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SSR4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SSR4
Gene Accession No. P51571
Gene Alias AW743063; C78062; CDG1Y; signal sequence receptor 4; signal sequence receptor subunit 4; signal sequence receptor subunit delta; signal sequence receptor, delta; Ssr4; SSR-delta; Translocon-associated protein subunit delta; Trap; TRAPD; TRAP-delta
Gene Symbols SSR4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SSR4. Recombinant protein control fragment (Product #RP-92340).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6748
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.