Learn More
Invitrogen™ SSRP1 Polyclonal Antibody

Description
Immunogen sequence: DDAEVSLMEV RFYVPPTQED GVDPVEAFAQ NVLSKADVIQ ATGDAICIFR ELQCLTPRGR YDIRIYPTFL HLHGKTFDYK IPYTTVLRLF LLPHKDQRQM FFVISLDPPI KQGQTRYHFL ILLFSKDEDI SLTLNMNEEE VE Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 100%.
Specifications
Specifications
| Antigen | SSRP1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | SSRP1 |
| Gene Accession No. | Q08945 |
| Gene Alias | C81323; chromatin-specific transcription elongation factor 80 kDa subunit; Ciidbp; cisplatin-DNA SSRP; facilitates chromatin remodeling 80 kDa subunit; facilitates chromatin transcription complex 80 kDa subunit; Facilitates chromatin transcription complex subunit SSRP1; FACT; FACT 80 kDa subunit; FACT complex subunit SSRP1; FACT80; FACTp80; high mobility group box; Hmg1-rs1; Hmgi-rs3; Hmgox; hSSRP1; recombination signal sequence recognition protein 1; Ssrp1; structure specific recognition protein 1; structure-specific recognition protein 1; T160 |
| Gene Symbols | SSRP1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.