Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SSRP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589337
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589337 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589337 Supplier Invitrogen™ Supplier No. PA589337
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAETLEFNDV YQEVKGSMND GRLRLSRQGI IFKNSKTGKV DNIQAGELTE GIWRRVALGH GLKLLTKNGH VYKYDGFRES EFEKLSDFFK THYRLELMEK DLCVKGWNWG TVKFGGQLLS FDIGDQPVFE IPLSNVSQCT TGKNEVTLEF HQNDDAEVSL MEVRFYVPPT QEDGVDPVEA FAQNVLSKAD VIQATGDAIC IFRELQCLTP RGRYDIRIYP TFLHLHGKTF DYKIPYTTVL RLFLLPHKDQ RQMFFVISLD PPIKQGQTRY HFLILLFSKD EDISLTLNMN EEEVEKRFEG; Positive Samples: SW620, HeLa; Cellular Location: Chromosome, Nucleus, nucleolus.

SPT16 and SSRP1 are found in all eurkaryotic cells and exists as part of an essential heterodimer complex. Together, SPT 16 and SSRP1 are a highly abundant nuclear complex that in mammals has been called FACT, or Facilitates Chromatin Transcription. FACT enhances the processivity of RNA polymerase II through nucleosomes. Evidence also points to a role for FACT in replication, possibly by enhancing DNA polymerase through nucleosomes as well.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SSRP1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.32 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SSRP1
Gene Accession No. Q08945
Gene Alias C81323; chromatin-specific transcription elongation factor 80 kDa subunit; Ciidbp; cisplatin-DNA SSRP; facilitates chromatin remodeling 80 kDa subunit; facilitates chromatin transcription complex 80 kDa subunit; Facilitates chromatin transcription complex subunit SSRP1; FACT; FACT 80 kDa subunit; FACT complex subunit SSRP1; FACT80; FACTp80; high mobility group box; Hmg1-rs1; Hmgi-rs3; Hmgox; hSSRP1; recombination signal sequence recognition protein 1; Ssrp1; structure specific recognition protein 1; structure-specific recognition protein 1; T160
Gene Symbols SSRP1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human SSRP1 (NP_0031371).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6749
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.